Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Pentylcyclohexanecarboxylic Acid (cis- and trans- mixture) 98.0+%, TCI America™
SDP

Catalog No. P20455G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P20455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P20455G Supplier TCI America Supplier No. P20455G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 38792-89-1
Molecular Formula C12H22O2
Molecular Weight (g/mol) 198.306
MDL Number MFCD00466238
InChI Key RVLAXPQGTRTHEV-UHFFFAOYSA-N
Synonym 4-Amylcyclohexanecarboxylic Acid
PubChem CID 98882
IUPAC Name 4-pentylcyclohexane-1-carboxylic acid
SMILES CCCCCC1CCC(CC1)C(=O)O

Specifications

Color Yellow
Quantity 5 g
Formula Weight 198.31
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Pentylcyclohexanecarboxylic Acid (cis- and trans- mixture)
EINECSNumber (3)-3723
RTECSNumber GU8622500
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.