Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Pentylcyclohexanone 95.0+%, TCI America™
SDP

Catalog No. P08835G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P08835G 5 g
P088325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P08835G Supplier TCI America Supplier No. P08835G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 61203-83-6
Molecular Formula C11H20O
Molecular Weight (g/mol) 168.28
MDL Number MFCD00059581
InChI Key UKLNPJDLSPMJMQ-UHFFFAOYSA-N
Synonym 4-pentylcyclohexanone, 4-n-pentylcyclohexanone, 4-amylcyclohexanone, 4-n-amylcyclohexanone, 4-pentyl-1-cyclohexanone, 4-pentylcyclo hexanone, pubchem20247, acmc-209mq3, uklnpjdlspmjmq-uhfffaoysa
PubChem CID 566097
IUPAC Name 4-pentylcyclohexan-1-one
SMILES CCCCCC1CCC(=O)CC1

Specifications

Color Yellow
Quantity 5 g
UN Number 3082
Formula Weight 168.28
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Pentylcyclohexanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.