Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Pentyloxyaniline, 99%, Thermo Scientific™

Catalog No. AC206660050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC206660050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC206660050 Supplier Thermo Scientific Chemicals Supplier No. 206660050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39905-50-5
Molecular Formula C11H17NO
Molecular Weight (g/mol) 179.26
MDL Number MFCD00007867
InChI Key QZLNSNIHXKQIIS-UHFFFAOYSA-N
Synonym 4-pentyloxyaniline, p-pentyloxyaniline, 4-pentyloxy aniline, benzenamine, 4-pentyloxy, 4-n-pentoxyaniline, 4-pentyloxyphenylamine, p-pentyloxy aniline, p-n-pentoxy aniline, 4-pentyloxy benzenamine, p-aminophenyl pentyl ether
PubChem CID 94500
IUPAC Name 4-(pentyloxy)aniline
SMILES CCCCCOC1=CC=C(N)C=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Brown-Yellow
Density 0.97g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Refractive Index 1.5310 to 1.5340 (20°C, 589nm)
Linear Formula CH3(CH2)4OC6H4NH2
Quantity 5 g
Beilstein 13, III, 998
Formula Weight 179.26
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material 4-Pentyloxyaniline, 99%
Show More Show Less
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Recommended Storage Product may darken during storage
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.