Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-Pentylphenol-d11 | 1219805-30-7 | 98.3% | 175.31 | 10 MG
SDP

Catalog No. 5000453511
Encompass_Preferred

4-Pentylphenol-d11 is a deuterium-labeled chemical compound with a molecular weight of 175.31. It appears as a colorless to light yellow liquid and is specified with a purity of 98.3%.

  • Deuterium-labeled compound
  • Purity of 98.3% (GC)
  • Colorless to light yellow liquid appearance
  • Isotopic enrichment of 98.4%
  • Storage conditions: -20°C for 3 years (pure form); -80°C for 6 months or -20°C for 1 month (in solvent)

Catalog No. 50-004-53511 Supplier Medchemexpress LLC Supplier No. HYW010112S110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Pentylphenol-d11 is a deuterium-labeled chemical compound with a molecular weight of 175.31. It appears as a colorless to light yellow liquid and is specified with a purity of 98.3%.

  • Deuterium-labeled compound
  • Purity of 98.3% (GC)
  • Colorless to light yellow liquid appearance
  • Isotopic enrichment of 98.4%
  • Storage conditions: -20°C for 3 years (pure form); -80°C for 6 months or -20°C for 1 month (in solvent)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.