Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Pentyn-1-ol

Catalog No. 30248125G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-248-125G 25 g
30-248-15G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 30-248-125G Supplier Sigma Aldrich Supplier No. 30248125G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Mechanism of (thf)w(co)5-promoted endo- and exo-cycloisomerization of 4-pentyn-1-ol was studied.4-pentyn-1-ol was used in preparation of 3-pent-4-ynyloxy phthalonitrile
  • It was also used as starting reagent in stereoselective total synthesis of antimicrobial marine metabolites, ieodomycin a and b

Specifications

Density 0.904 g/mL (at 25°C (literature))
Boiling Point 154°C to 155°C (lit.)
Molecular Formula C5H8O
Refractive Index n20/D 1.445 (literature)
Linear Formula HC///CCH2CH2CH2OH
MDL Number MFCD00002974
Quantity 25 g
Molecular Weight (g/mol) 84.12
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.