Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Pentyn-1-ol 96.0+%, TCI America™
SDP

Catalog No. P081725ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P081725ML 25 mL
P08175ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P081725ML Supplier TCI America Supplier No. P081725ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5390-04-5
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.12
MDL Number MFCD00002974
InChI Key CRWVOXFUXPYTRK-UHFFFAOYSA-N
Synonym 4-pentyn-1-ol, 4-pentyne-1-ol, 1-pentyn-5-ol, 5-hydroxypentyne, 4-pentynol, 4-pentin-1-ol, pent-4yn-1-ol, 5-hydroxy-1-pentyne, pent-1-yn-5-ol, pubchem2025
PubChem CID 79346
IUPAC Name pent-4-yn-1-ol
SMILES C#CCCCO

Specifications

Color Yellow
Boiling Point 155°C
Quantity 25 mL
Formula Weight 84.12
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Pentyn-1-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.