Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-pentynoic acid, 2,5-dioxo-1-pyrrolidinyl ester | 132178-37-1 | 99.8% | 195.17 g/mol | C9H9NO4 | 1 G
SDP

Catalog No. 5000202775
Encompass_Preferred

Propargyl-C1-NHS ester is an alkyne-containing N-hydroxysuccinimide (NHS) ester used as a non-cleavable, amine-reactive linker for bioconjugation. It reacts with primary amines to form stable amide bonds and participates in copper-catalyzed azide-alkyne cycloaddition (CuAAC) for click chemistry applications.

  • Alkyne-containing N-hydroxysuccinimide ester reactive toward primary amines.
  • Copper-catalyzed azide-alkyne cycloaddition (CuAAC) for click chemistry.
  • Forms stable amide bonds with amine-containing biomolecules.
  • Suitable for antibody-drug conjugation and general bioconjugation workflows.
  • Recommended storage: -20°C, sealed and protected from moisture and light.
  • Molecular weight 195.17 g/mol; formula C9H9NO4; cas number 132178-37-1.

Catalog No. 50-002-02775 Supplier Medchemexpress LLC Supplier No. HY1265111G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propargyl-C1-NHS ester is an alkyne-containing N-hydroxysuccinimide (NHS) ester used as a non-cleavable, amine-reactive linker for bioconjugation. It reacts with primary amines to form stable amide bonds and participates in copper-catalyzed azide-alkyne cycloaddition (CuAAC) for click chemistry applications.

  • Alkyne-containing N-hydroxysuccinimide ester reactive toward primary amines.
  • Copper-catalyzed azide-alkyne cycloaddition (CuAAC) for click chemistry.
  • Forms stable amide bonds with amine-containing biomolecules.
  • Suitable for antibody-drug conjugation and general bioconjugation workflows.
  • Recommended storage: -20°C, sealed and protected from moisture and light.
  • Molecular weight 195.17 g/mol; formula C9H9NO4; cas number 132178-37-1.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.