Learn More
Medchemexpress LLC 4-pentynoic acid | 6089-09-4 | MFCD00004407 | 98.4% | 98.10 | C5H6O2 | 10g
SDP
Catalog No. 5000266660
Encompass_Preferred
4-Pentynoic acid (Propargylacetic acid) is an intermediate to produce biologically active compounds 4-Pentynoic acid is widely utilized as a building block for the synthesis of eight sequence-defined model oligomers[1] 4-Pentynoic acid is a click chemistry reagent it contains an Alkyne group and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups
Catalog No. 50-002-66660 Supplier Medchemexpress LLC Supplier No. HYY123010G
4-Pentynoic acid (Propargylacetic acid) is an intermediate to produce biologically active compounds 4-Pentynoic acid is widely utilized as a building block for the synthesis of eight sequence-defined model oligomers[1] 4-Pentynoic acid is a click chemistry reagent it contains an Alkyne group and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups