Learn More
Ready America™ 4 Person 3 Day Elite Emergency Backpack
SDP
Catalog No. 17467155
Change view
Click to view available options
Color:
Black
Catalog No. Color
17-467-155 Black
1 options
Catalog No. 17-467-155 Supplier Ready America™ Supplier No. 70452
Add to Cart
Add to Cart
Elite 4-Person Emergency backpack has all the emergency essentials for 3 days (72 hours) after a disaster.
  • The Emergency Power Station is a four-function hand-crank unit that includes a flashlight, AM/FM radio, cell phone charger and a siren. One minute of cranking produces either 30 minutes of bright light, 2 minutes of talk time on a cell phone, or 15 minutes of radio. The Heavy Duty Multi-Tool includes: knife, screwdriver, bottle opener, saw, fire starter and pliers.
  • Emergency Kit Contains:
    • (6) Food Bar (2400 Calorie) (5-year shelf life)
    • (36) Pouches of Water (4.225 oz.) (5-year shelf life)
    • (2) 2 Person Tube Tent
    • (1) Folding Shovel
    • (1) Heavy Duty Work Gloves
    • (4) Knit Cap (Beanie)
    • (4) Niosh N-95 Approved Dust Mask
    • (8) 10 in. Nitrile Gloves
    • (2) Hygiene Kit Basic
    • (2) Extra Toothbrush
    • (2) Extra Mouthwash
    • (1) Potable Aqua Germicidal Tablets (50 count)
    • (3) Bio Hazard Bags
    • (1) First-Aid Kit (107 piece)
    • (1) Heavy Duty Multi-Tool
    • (1) 8 in. Survival Knife with Compass & Survival Kit (matches, needle/thread, fishing line/hook)
    • (1) Duct Tape (10 yards)
    • (1) 5 in 1 Survival Whistle
    • (4) Emergency Poncho
    • (4) Emergency Survival Blanket
    • (4) Pocket Tissue
    • (12) Wet Naps
    • (1) Wipes in Soft Dispenser
    • (1) Micro Spark Wheel
    • (1) Heritage Light-Me Tinder (12 pack)
    • (1) Insta-Fire (3 pack)
    • (1) Deck of Playing Cards
    • (1) Emergency Power Station 4 Function
    • (4) 12 Hour Light Stick
    • (1) Water Bottle (1 Liter)
    • (1) Filtered Water Bottle
    • (1) Hand Sanitizer (4 oz.)
    • (1) 25 ft. Cord
    • (1) Document Pouch
    • (1) X-Large Elite Backpack
Specifications
Product Type Emergency Backpack
Color Black
Disposable Reusable