Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ready America™ 4 Person Deluxe Emergency Kit Bucket
SDP

Catalog No. 19777017
Change view
Click to view available options
Color:
Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-777-017 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-777-017 Supplier Ready America™ Supplier No. 70395
  • $314.00
    $241.65 / Each of 1

    Save $72.35

    23% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ready America™ 4 Person Deluxe Emergency Kit Bucket allows a group of up to four people to be prepared when spending long periods of time together.

  • Inside of this carefully-stocked, secure and waterproof bucket is enough packaged food and water to sustain a family of four for the critical first 72 hours of an emergency situation
  • Includes the following:
    • 4 Food Bars 2400 Calories
    • 4 One Liter Boxes of Water
    • 4 Emergency Ponchos
    • 4 Survival Blankets
    • 4 Emergency Lightsticks (12 Hour)
    • 4 Pairs Nitrile Gloves
    • 4 Niosh N-95 Dust Masks
    • 4 Pocket Tissue Packs
    • 1 Emergency Whistle
    • 1 Pair Leather Work Gloves
    • 1 Multi-Function Tool
    • 1 Duct Tape (10 yrds)
    • 4 Safety Goggles
    • 3 Bio-Hazard Bags
    • 12 Pre-Moistened Towelettes
    • 1 First Aid Kit (107 Pieces)
    • 1 Emergency Power Station: Flashlight / AM-FM Radio / Siren / Cell Phone Charger
    • A 5-Gallon Bucket
    • 1 Bucket Lid

Specifications

Product Type Emergency Kit
Color Red
Material Plastic
Height (English) 15 in.
Height (Metric) 38.1 cm
Length (English) 12 in.
Length (Metric) 30.48 cm
Weight (English) 24 lb.
Weight (Metric) 10.9 kg
Width (English) 12 in.
Width (Metric) 30.48 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.