Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ready America™ 4 Person Deluxe Emergency Kit
SDP

Catalog No. 19777016
Change view
Click to view available options
Color:
Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-777-016 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-777-016 Supplier Ready America™ Supplier No. 70385
  • $353.00
    $271.65 / Each of 1

    Save $81.35

    23% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ready America™ 4 Person Deluxe Emergency Kit is designed to lasts up to 3 days and includes unique items such as an emergency power station.

  • Includes the following:
    • 4 Food Bars (2400 Calorie) (5-year shelf life)
    • 4 Boxes of Water (1 Liter) (5-year shelf life)
    • 4 Emergency Ponchos
    • 4 Survival Blankets
    • 1 Emergency Power Station: Flashlight/AM-FM Radio/Siren/Phone Charger
    • 1 Duct Tape (10 yards)
    • 1 Multi-Function Pocket Tool
    • 1 First-Aid Kit (107 piece)
    • 4 Emergency Lightsticks (12-hr)
    • 8 10 in. Nitrile Gloves
    • 4 Safety Goggles
    • 4 Niosh N-95 Dust Mask
    • 4 Pocket Tissue Packs
    • 1 Pair of Leather Gloves
    • 1 Emergency Whistle
    • 12 Pre-Moistened Towelettes
    • 3 Bio-Hazard Bags
    • 1 Backpack

Specifications

Product Type Emergency Kit
Color Red
Material Canvas
Height (English) 10 in.
Height (Metric) 25.4 cm
Length (English) 14 in.
Length (Metric) 35.56 cm
Weight (English) 21 lb.
Weight (Metric) 9.54 kg
Width (English) 10 in.
Width (Metric) 25.4 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.