Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Phenoxyacetophenone, TCI America™
SDP

Catalog No. P13555G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P13555G Supplier TCI America Supplier No. P13555G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5031-78-7
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.25
MDL Number MFCD00008744
InChI Key DJNIFZYQFLFGDT-UHFFFAOYSA-N
Synonym 4'-phenoxyacetophenone, 1-4-phenoxyphenyl ethanone, 4-phenoxyacetophenone, p-phenoxyacetophenone, 1-4-phenoxyphenyl ethan-1-one, ethanone, 1-4-phenoxyphenyl, 4-acetyldiphenyl ether, 1-4-phenoxyphenyl-1-ethanone, 1-4-phenoxyphenyl ethanon, pubchem13195
PubChem CID 236783
IUPAC Name 1-(4-phenoxyphenyl)ethan-1-one
SMILES CC(=O)C1=CC=C(OC2=CC=CC=C2)C=C1

Specifications

Melting Point 50°C
Color White
Boiling Point 325°C
Quantity 5 g
Formula Weight 212.25
Physical Form Crystalline Powder
Chemical Name or Material 4′-Phenoxyacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.