Learn More
Medchemexpress LLC 4-Phenoxybenzoic acid | 2215-77-2 | 2539 | 99.8% | C13H10O3 | 25 G
SDP
Catalog No. 5000358938
Encompass_Preferred
4-Phenoxybenzoic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It has been shown to block DNA binding of the human papillomarvirus (HPV) E2 protein. Additionally, it serves as an intermediate in the synthesis of Sitafloxacin, a fluoroquinolone antibiotic that shows potential in treating Buruli ulcer.
  • Blocks DNA binding of the human papillomarvirus (HPV) E2 protein in vitro
  • Employed as an intermediate of sitafloxacin, a fluoroquinolone antibiotic
Catalog No. 50-003-58938 Supplier Medchemexpress LLC Supplier No. 25GHY78854
4-Phenoxybenzoic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It has been shown to block DNA binding of the human papillomarvirus (HPV) E2 protein. Additionally, it serves as an intermediate in the synthesis of Sitafloxacin, a fluoroquinolone antibiotic that shows potential in treating Buruli ulcer.
  • Blocks DNA binding of the human papillomarvirus (HPV) E2 protein in vitro
  • Employed as an intermediate of sitafloxacin, a fluoroquinolone antibiotic