Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Phenoxybenzoic acid, 99%, Thermo Scientific Chemicals

Catalog No. AAA1799814
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1799814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1799814 Supplier Thermo Scientific Chemicals Supplier No. A1799814
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2215-77-2 | Molecular Formula: C13H10O3 | Molecular Weight: 214.22 g/mol

4-Phenoxybenzoic acid can block DNA binding of the human papillomarvirus (HPV) E2 protein. It is employed as an intermediate of Sitafloxacin, which is a fluoroquinolone antibiotic that shows promise in the treatment of Buruli ulcer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Phenoxybenzoic acid can block DNA binding of the human papillomarvirus (HPV) E2 protein. It is employed as an intermediate of Sitafloxacin, which is a fluoroquinolone antibiotic that shows promise in the treatment of Buruli ulcer.

Solubility
Very faint turbidity in hot methanol.

Notes
Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2215-77-2
Molecular Formula C13H10O3
Molecular Weight (g/mol) 214.22
MDL Number MFCD00002539
InChI Key RYAQFHLUEMJOMF-UHFFFAOYSA-N
Synonym p-phenoxybenzoic acid, benzoic acid, 4-phenoxy, diphenyl ether 4-carboxylic acid, 4-phenoxy-benzoic acid, 4-carboxybiphenyl ether, 4-carboxydiphenyl ether, benzoic acid, p-phenoxy, p-carboxydiphenyl ether, 4-phenoxy benzoic acid, 4-phoc6h4cooh
PubChem CID 75182
ChEBI CHEBI:72632
IUPAC Name 4-phenoxybenzoic acid
SMILES OC(=O)C1=CC=C(OC2=CC=CC=C2)C=C1

Specifications

Melting Point 160°C to 164°C
Quantity 25 g
Beilstein 2212463
Solubility Information Very faint turbidity in hot methanol.
Formula Weight 214.22
Percent Purity 99%
Chemical Name or Material 4-Phenoxybenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 218-682-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.