Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-phenoxybenzoyl chloride, 97%, Thermo Scientific™

Catalog No. CC53702CB
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC53702CB 250 mg Amber Glass Bottle
CC53702DA 1 g Amber Glass Bottle
CC53702DE 5 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC53702CB Supplier Thermo Scientific Chemicals Supplier No. CC53702CB

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1623-95-6
Molecular Formula C13H9ClO2
Molecular Weight (g/mol) 232.663
InChI Key AOOZVQGGMFGGEE-UHFFFAOYSA-N
PubChem CID 74193
IUPAC Name 4-phenoxybenzoyl chloride
SMILES C1=CC=C(C=C1)OC2=CC=C(C=C2)C(=O)Cl

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym benzoyl chloride, 4-phenoxy, p-phenoxybenzoyl chloride, 4-phenoxybenzoylchloride, benzoyl chloride,4-phenoxy, p-phenoxybenzoylchloride, 4-phenoxy benzoyl chloride, 4-phenoxy-benzoyl chloride, 4-phenoxybenzoyl chlo-ride, benzoyl chloride, p-phenoxy, 4-phenoxyphenylcarbonyl chloride
Formula Weight 232.67
Percent Purity 97%
Chemical Name or Material 4-Phenoxybenzoyl chloride
Hazard Statement GHS P Statement
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.