Learn More
Medchemexpress LLC HY-18563 100mg Medchemexpress, 4-Phenoxybenzylamine CAS:107622-80-0 Purity:>98%
SDP
Catalog No. 502082046
Encompass_Preferred
Medchemexpress, HY-18563 100mg 4-Phenoxybenzylamine CAS:107622-80-0 4-Phenoxybenzylamine inhibits the function of the NS3 protein by stabilizing an inactive conformation with an IC50 of about 500 μM against FL NS3/4a. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2046 Supplier Medchemexpress LLC Supplier No. HY18563100MG
Medchemexpress, HY-18563 100mg 4-Phenoxybenzylamine CAS:107622-80-0 4-Phenoxybenzylamine inhibits the function of the NS3 protein by stabilizing an inactive conformation with an IC50 of about 500 μM against FL NS3/4a. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.