Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Phenoxyphenyl isocyanate, 98%

Catalog No. aal1168603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1168603 1 g
AAL1168606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1168603 Supplier Thermo Scientific Chemicals Supplier No. L1168603
Only null left
Add to Cart

CAS: 59377-19-4 | C13H9NO2 | 211.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59377-19-4
Molecular Formula C13H9NO2
Molecular Weight (g/mol) 211.22
MDL Number MFCD00013876
InChI Key PNBUGOFIKAHZRW-UHFFFAOYSA-N
Synonym 4-phenoxyphenyl isocyanate, 4-phenoxyphenylisocyanate, 1-isocyanato-4-phenoxy-benzene, benzene, 1-isocyanato-4-phenoxy, acmc-20ambk, pubchem17221, p-phenoxyphenylisocyanate, 4-phenoxybenzenisocyanate, 4-phenoxyphenyl-isocyanate, 4-isocyanatodiphenyl ether
PubChem CID 2734896
IUPAC Name 1-isocyanato-4-phenoxybenzene
SMILES O=C=NC1=CC=C(OC2=CC=CC=C2)C=C1

Specifications

Density 1.169
Boiling Point 169°C to 170°C (3 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.595
Quantity 1 g
UN Number UN2206
Beilstein 393196
Sensitivity Moisture sensitive
Formula Weight 211.22
Percent Purity 98%
Chemical Name or Material 4-Phenoxyphenyl isocyanate
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.