Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Phenoxyphenylacetonitrile, 98%, Thermo Scientific™

Catalog No. AAL1057203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1057203 1 g
AAL1057206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1057203 Supplier Thermo Scientific Chemicals Supplier No. L1057203

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Phenoxyphenylacetonitrile is involved in the preparation of 2-(4-phenoxybenzoyl)-5-hydroxyindole, which is used as aCa2+/calmodulin-dependent protein kinase II (CaMKII) inhibitor.

Solubility
Miscible with toluene.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92163-15-0
Molecular Formula C14H11NO
Molecular Weight (g/mol) 209.248
MDL Number MFCD00016399
InChI Key PCOVIPKFOQOPEH-UHFFFAOYSA-N
Synonym 4-phenoxyphenylacetonitrile, 2-4-phenoxyphenyl acetonitrile, 4-phenoxybenzyl cyanide, 2-4-phenoxyphenyl ethanenitrile, timtec-bb sbb005900, 4-phenoxylphenylacetonitrile, 4-phenoxy-benzeneacetonitrile, benzeneacetonitrile, 4-phenoxy, p-phenoxyphenylacetonitrile
PubChem CID 2777953
IUPAC Name 2-(4-phenoxyphenyl)acetonitrile
SMILES C1=CC=C(C=C1)OC2=CC=C(C=C2)CC#N

Specifications

Density 1.12
Boiling Point 145°C (1 mmHg)
Refractive Index 1.582
Quantity 1 g
Beilstein 2693571
Solubility Information Miscible with toluene.
Formula Weight 209.25
Percent Purity 98%
Chemical Name or Material 4-Phenoxyphenylacetonitrile
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.