Learn More
Medchemexpress LLC 4-Phenyl-1,2,4-triazoline-3,5-dione | 4233-33-4 | MFCD00003148 | 98.0% | 175.14 g/mol | C8H5N3O2 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW016748
Encompass_Preferred
PTAD (4-Phenyl-1,2,4-triazoline-3,5-dione) is a highly reactive dieneophile used in organic synthesis. It participates in rapid Diels-Alder reactions and can oxidize thiols to disulfides, making it useful for synthetic method development and research applications.
  • Highly reactive dieneophile enabling rapid Diels-Alder cycloadditions.
  • Oxidizes thiols to disulfides selectively under mild conditions.
  • High purity suitable for research applications.
  • Solid-state red to dark-red powder, easy to handle and store.
  • Identified by CAS 4233-33-4 for unambiguous reference.
  • Useful in synthetic method development and mechanistic studies.
Catalog No. 50-003-03820