Learn More
Medchemexpress LLC 4-phenyl-1-(4-phenylbutyl)piperidine maleate | 201216-39-9 | MFCD00209909 | 99.6% | 409.52 g/mol | C25H31NO4 | 5 MG
SDP
Catalog No. 5000212030
Encompass_Preferred
4-PPBP maleate is a research-grade sigma-1 (σ1) receptor ligand and agonist supplied as the maleate salt in powder form. It is used in neuroscience and pharmacology research to probe receptor activity, intracellular signaling, and neuroprotective mechanisms. Consult the product documentation for handling, storage, and purity details.
  • Potent sigma-1 receptor ligand and agonist suitable for receptor pharmacology studies.
  • High reported purity for consistent experimental results.
  • Provided as a stable maleate salt in powder form for storage and formulation flexibility.
  • Available in small pack sizes appropriate for research use.
  • Documentation available for handling and quality control, including SDS and COA.
Catalog No. 50-002-12030 Supplier Medchemexpress LLC Supplier No. HY1010435MG
4-PPBP maleate is a research-grade sigma-1 (σ1) receptor ligand and agonist supplied as the maleate salt in powder form. It is used in neuroscience and pharmacology research to probe receptor activity, intracellular signaling, and neuroprotective mechanisms. Consult the product documentation for handling, storage, and purity details.
  • Potent sigma-1 receptor ligand and agonist suitable for receptor pharmacology studies.
  • High reported purity for consistent experimental results.
  • Provided as a stable maleate salt in powder form for storage and formulation flexibility.
  • Available in small pack sizes appropriate for research use.
  • Documentation available for handling and quality control, including SDS and COA.