Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Phenyl-1-cyclohexene 96.0+%, TCI America™
SDP

Catalog No. P14451G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P14451G 1 g
P14455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P14451G Supplier TCI America Supplier No. P14451G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4994-16-5
Molecular Formula C12H14
Molecular Weight (g/mol) 158.244
MDL Number MFCD00049188
InChI Key XWCWNUSFQVJNDI-UHFFFAOYSA-N
Synonym 3-Cyclohexen-1-ylbenzene
PubChem CID 21096
IUPAC Name cyclohex-3-en-1-ylbenzene
SMILES C1CC(CC=C1)C2=CC=CC=C2

Specifications

Color Yellow
Boiling Point 235°C
Quantity 1 g
Formula Weight 158.24
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Phenyl-1-cyclohexene
RTECSNumber CZ1301000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.