Learn More
Medchemexpress LLC 4-phenyl-2-butanone | 2550-26-7 | MFCD00008790 | 99.8% | 148.21 g/mol | C10H12O | 10 G
SDP
Catalog No. 5000300321
Encompass_Preferred
Benzylacetone (2-butanone, 4-phenyl-) is an aromatic ketone used as a biochemical reagent and reported tyrosinase inhibitor. It is supplied as a colorless to light yellow liquid with high reported purity (99.78%), molecular formula C10H12O, and molecular weight 148.21 g/mol. Common uses include fragrance intermediate work and enzymatic research.
Catalog No. 50-003-00321 Supplier Medchemexpress LLC Supplier No. HYW01561610G
Benzylacetone (2-butanone, 4-phenyl-) is an aromatic ketone used as a biochemical reagent and reported tyrosinase inhibitor. It is supplied as a colorless to light yellow liquid with high reported purity (99.78%), molecular formula C10H12O, and molecular weight 148.21 g/mol. Common uses include fragrance intermediate work and enzymatic research.