Learn More
Medchemexpress LLC 4-phenyl-2-butanone | 2550-26-7 | MFCD00008790 | 99.8% | 148.21 g·mol⁻¹ | C10H12O | 100 G
SDP
Catalog No. 5000300733
Encompass_Preferred
Benzylacetone (4-phenyl-2-butanone) is an aromatic ketone used as a research reagent with reported biological activities, including reversible inhibition of tyrosinase and effects on appetite and locomotion. It is supplied at high purity, is soluble in DMSO, and has defined storage recommendations for both the neat material and solutions.
  • High purity (99.78%) suitable for biochemical assays.
  • Reversible tyrosinase inhibitor useful in enzyme studies.
  • Soluble in DMSO for easy assay formulation.
  • Stable under recommended storage conditions.
  • Known CAS number 2550-26-7 for unambiguous identification.
Catalog No. 50-003-00733 Supplier Medchemexpress LLC Supplier No. HYW015616100G
Benzylacetone (4-phenyl-2-butanone) is an aromatic ketone used as a research reagent with reported biological activities, including reversible inhibition of tyrosinase and effects on appetite and locomotion. It is supplied at high purity, is soluble in DMSO, and has defined storage recommendations for both the neat material and solutions.
  • High purity (99.78%) suitable for biochemical assays.
  • Reversible tyrosinase inhibitor useful in enzyme studies.
  • Soluble in DMSO for easy assay formulation.
  • Stable under recommended storage conditions.
  • Known CAS number 2550-26-7 for unambiguous identification.