Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-phenyl-2-pyrrolidinone | 1198-97-6 | MFCD01687226 | >98.0% | 161.20 | C10H11NO | 25 G
SDP

Catalog No. 5000302804
Encompass_Preferred

4-Phenyl-2-pyrrolidinone (CAS 1198-97-6) is a chemical intermediate and precursor used in medicinal chemistry research and drug synthesis. It is employed as a building block for preparing pharmacologically active compounds, with reported use in the synthesis of anticonvulsant and nootropic analogs.

  • Purity >98.0% (GC)
  • Molecular formula C10H11NO; molecular weight 161.20
  • Functions as a drug intermediate and synthetic precursor
  • Offered in laboratory pack sizes from 5 g to 100 g
  • White to pale yellow powder appearance suitable for research handling

Catalog No. 50-003-02804 Supplier Medchemexpress LLC Supplier No. 25GHYW017851
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Phenyl-2-pyrrolidinone (CAS 1198-97-6) is a chemical intermediate and precursor used in medicinal chemistry research and drug synthesis. It is employed as a building block for preparing pharmacologically active compounds, with reported use in the synthesis of anticonvulsant and nootropic analogs.

  • Purity >98.0% (GC)
  • Molecular formula C10H11NO; molecular weight 161.20
  • Functions as a drug intermediate and synthetic precursor
  • Offered in laboratory pack sizes from 5 g to 100 g
  • White to pale yellow powder appearance suitable for research handling

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.