Learn More
Medchemexpress LLC 4-phenyl-2-pyrrolidinone | 1198-97-6 | MFCD01687226 | >98.0% | 161.20 g/mol | C10H11NO | 5 G
SDP
Catalog No. 5000302041
Encompass_Preferred
4-Phenyl-2-pyrrolidinone (CAS 1198-97-6) is a small-molecule synthetic intermediate and precursor used in organic synthesis and medicinal chemistry research. It is supplied as a solid for research and development applications and is reported in literature as an intermediate in the synthesis of anticonvulsant and nootropic compounds.
  • High purity (≥98.0%) as stated on certificate of analysis.
  • White to off-white solid appearance.
  • Molecular formula C10H11NO; molecular weight 161.20 g/mol.
  • Suitable as a precursor and synthetic intermediate in medicinal chemistry.
  • Packaged for small-scale research use.
Catalog No. 50-003-02041 Supplier Medchemexpress LLC Supplier No. HYW0178515G
4-Phenyl-2-pyrrolidinone (CAS 1198-97-6) is a small-molecule synthetic intermediate and precursor used in organic synthesis and medicinal chemistry research. It is supplied as a solid for research and development applications and is reported in literature as an intermediate in the synthesis of anticonvulsant and nootropic compounds.
  • High purity (≥98.0%) as stated on certificate of analysis.
  • White to off-white solid appearance.
  • Molecular formula C10H11NO; molecular weight 161.20 g/mol.
  • Suitable as a precursor and synthetic intermediate in medicinal chemistry.
  • Packaged for small-scale research use.