Learn More
Medchemexpress LLC 4-phenyl-2-pyrrolidinone | 1198-97-6 | MFCD01687226 | 99.9% | 161.20 g/mol | C10H11NO | 100 G
SDP
Catalog No. 5000299630
Encompass_Preferred
4-Phenyl-2-pyrrolidinone is a chemical intermediate and precursor used in medicinal chemistry and organic synthesis. Supplied as a solid for research use only, it is commonly employed in the synthesis of anticonvulsant and nootropic compounds and as an intermediate in the preparation of BRD4 bromodomain inhibitors.
  • High purity (99.93%) suitable for research applications.
  • Solid, white to off-white appearance for easy handling.
  • Available in multiple pack sizes to support small-scale and bulk synthesis.
  • Stable under recommended storage conditions for reliable shelf life.
  • Intended for use as a precursor and synthetic intermediate in medicinal chemistry.
Catalog No. 50-002-99630 Supplier Medchemexpress LLC Supplier No. HYW017851100G
4-Phenyl-2-pyrrolidinone is a chemical intermediate and precursor used in medicinal chemistry and organic synthesis. Supplied as a solid for research use only, it is commonly employed in the synthesis of anticonvulsant and nootropic compounds and as an intermediate in the preparation of BRD4 bromodomain inhibitors.
  • High purity (99.93%) suitable for research applications.
  • Solid, white to off-white appearance for easy handling.
  • Available in multiple pack sizes to support small-scale and bulk synthesis.
  • Stable under recommended storage conditions for reliable shelf life.
  • Intended for use as a precursor and synthetic intermediate in medicinal chemistry.