Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Phenyl-5-(trifluoromethyl)thiophene-2-carboxylic acid, 95%, Thermo Scientific™

Catalog No. SEW02660DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SEW02660DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SEW02660DA Supplier Thermo Scientific Chemicals Supplier No. SEW02660DA
Add to Cart

Chemical Identifiers

CAS 208108-76-3
Molecular Formula C12H7F3O2S
Molecular Weight (g/mol) 272.241
InChI Key GIUAGKGTDDIMHB-UHFFFAOYSA-N
Synonym 4-phenyl-5-trifluoromethyl thiophene-2-carboxylic acid, 4-phenyl-5-trifluoromethyl-2-thiophenecarboxylic acid, 2-thiophenecarboxylic acid, 4-phenyl-5-trifluoromethyl, 2-thiophenecarboxylicacid, 4-phenyl-5-trifluoromethyl, maybridge3_006560, 4-phenyl-5-trifluoromethyl-thiophene-2-carboxylic acid, 4-phenyl-5-trifluoromethyl-2-thiophene carboxylic acid
PubChem CID 2783372
IUPAC Name 4-phenyl-5-(trifluoromethyl)thiophene-2-carboxylic acid
SMILES C1=CC=C(C=C1)C2=C(SC(=C2)C(=O)O)C(F)(F)F

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 272.25
Percent Purity 95%
Chemical Name or Material 4-Phenyl-5-(trifluoromethyl)thiophene-2-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.