Learn More
TARGETMOL CHEMICALS INC Research Chemical 100mg
SDP
Catalog No. 503038154
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 500 mg, 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 4-Phenyl-7,8-dihydroxycoumarin (7,8-Dihydroxy-4-phenylcoumarin) inhibits the activity of Staphylococcus aureus DNA helicasepurity: 99%
Catalog No. 50-303-8154 Supplier TARGETMOL CHEMICALS INC Supplier No. TJS0368100MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 500 mg, 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 4-Phenyl-7,8-dihydroxycoumarin (7,8-Dihydroxy-4-phenylcoumarin) inhibits the activity of Staphylococcus aureus DNA helicasepurity: 99%