Learn More
Medchemexpress LLC 4-Phenyl-7,8-dihydroxycoumarin | 842-01-3 | 99.35% | 50 MG
SDP
Catalog No. 5000342619
Encompass_Preferred
4-Phenyl-7,8-dihydroxycoumarin is a coumarin derivative used in bronchiectasis research. This product is strictly for research purposes and not for sale to patients.
  • Suitable for bronchiectasis research
  • Powder storage: -20°C for 3 years, or 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, or -20°C for 1 month
Catalog No. 50-003-42619 Supplier Medchemexpress LLC Supplier No. HY12841050MG
4-Phenyl-7,8-dihydroxycoumarin is a coumarin derivative used in bronchiectasis research. This product is strictly for research purposes and not for sale to patients.
  • Suitable for bronchiectasis research
  • Powder storage: -20°C for 3 years, or 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, or -20°C for 1 month