Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-Phenyl-7,8-dihydroxycoumarin | 842-01-3 | 99.4% | 10 MG
SDP

Catalog No. 5000339756
Encompass_Preferred

4-Phenyl-7,8-dihydroxycoumarin is a coumarin derivative for research use only, and can be used for bronchiectasis research.

  • Chemical formula: C15H10O4
  • Molecular weight: 254.24
  • Appearance: Solid, off-white to yellow
  • Solubility: 100 mg/mL in DMSO
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • Related disease areas: Neurological, Eye or Ear Disease, Inflammation or Immune System Disease, Neurodegenerative Disease, Parkinson's Disease
  • Signaling pathways: Inflammation/Immunology, Metabolic Enzyme/Protease, Neuronal Signaling, COMT

Catalog No. 50-003-39756 Supplier Medchemexpress LLC Supplier No. HY12841010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Phenyl-7,8-dihydroxycoumarin is a coumarin derivative for research use only, and can be used for bronchiectasis research.

  • Chemical formula: C15H10O4
  • Molecular weight: 254.24
  • Appearance: Solid, off-white to yellow
  • Solubility: 100 mg/mL in DMSO
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • Related disease areas: Neurological, Eye or Ear Disease, Inflammation or Immune System Disease, Neurodegenerative Disease, Parkinson's Disease
  • Signaling pathways: Inflammation/Immunology, Metabolic Enzyme/Protease, Neuronal Signaling, COMT

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.