Learn More
Medchemexpress LLC 4-Phenyl-7,8-dihydroxycoumarin | 842-01-3 | 99.4% | 100 MG
SDP
Catalog No. 5000339986
Encompass_Preferred
4-Phenyl-7,8-dihydroxycoumarin is a coumarin derivative that can be used for bronchiectasis research.
  • Can be used for bronchiectasis research
  • Exhibits cytotoxicity against human A375 cells (IC50: 39 μM) and human MRC5 cells (IC50: 47 μM)
  • Recommended storage temperature for powder is -20°C for 3 years and 4°C for 2 years
  • Recommended storage temperature for in solvent is -80°C for 6 months and -20°C for 1 month
Catalog No. 50-003-39986 Supplier Medchemexpress LLC Supplier No. HY128410100MG
4-Phenyl-7,8-dihydroxycoumarin is a coumarin derivative that can be used for bronchiectasis research.
  • Can be used for bronchiectasis research
  • Exhibits cytotoxicity against human A375 cells (IC50: 39 μM) and human MRC5 cells (IC50: 47 μM)
  • Recommended storage temperature for powder is -20°C for 3 years and 4°C for 2 years
  • Recommended storage temperature for in solvent is -80°C for 6 months and -20°C for 1 month