Learn More
Medchemexpress LLC 4-phenylbutan-2-one | 2550-26-7 | MFCD00008790 | 99.8% | 148.21 g/mol | C10H12O | 25 G
SDP
Catalog No. 5000304138
Encompass_Preferred
Benzylacetone (IUPAC: 4-phenylbutan-2-one, CAS 2550-26-7) is an aromatic ketone used as a research chemical, fragrance intermediate, and biochemical probe. It is a colorless to pale-yellow liquid with a sweet, floral odor and is supplied with high purity for laboratory applications.
  • High purity suitable for research and analytical use.
  • Molecular weight 148.21 g/mol (C10H12O).
  • Stable when stored under recommended conditions.
  • Used as a fragrance intermediate and tyrosinase inhibitor probe.
  • Available in laboratory pack sizes for small-scale experiments.
Catalog No. 50-003-04138 Supplier Medchemexpress LLC Supplier No. 25GHYW015616
Benzylacetone (IUPAC: 4-phenylbutan-2-one, CAS 2550-26-7) is an aromatic ketone used as a research chemical, fragrance intermediate, and biochemical probe. It is a colorless to pale-yellow liquid with a sweet, floral odor and is supplied with high purity for laboratory applications.
  • High purity suitable for research and analytical use.
  • Molecular weight 148.21 g/mol (C10H12O).
  • Stable when stored under recommended conditions.
  • Used as a fragrance intermediate and tyrosinase inhibitor probe.
  • Available in laboratory pack sizes for small-scale experiments.