Learn More
Apexbio Technology LLC 4-Phenylbutyric acid 1821-12-1 500mg
SDP
Supplier:  Apexbio Technology LLC C6831500
Encompass_Preferred
4-Phenylbutyric acid (1821-12-1) is a small-molecule modulator targeting cellular stress response pathways It is designed to act as a chemical chaperone facilitating protein folding and trafficking thereby regulating protein homeostasis and inflammatory signaling 4-Phenylbutyric acid exerts its biological activity primarily through modulation of cellular stress and inhibition of inflammatory signaling pathways In in vitro and in vivo studies 4-Phenylbutyric acid demonstrates anti-inflammatory activity with reported IC50 values typically ranging from 0 5 mM to 5 mM depending on the experimental system and cell type Based on these pharmacological properties 4-Phenylbutyric acid holds research potential in metabolic disease models genetic protein-misfolding disorders neurological dysfunction and urea cycle disturbances
Catalog No. 50-000-90110