Learn More
Cayman Chemical 4 PhenylcarbonylbenzoIc AcI 1g
SDP
Catalog No. 502991204
Encompass_Preferred
A photooxidant; upon light activation, it forms an electrophilic aromatic ketone that acts as an oxidant in organic synthesis or biological systems; has been used in the study of amino acid oxidation, as well as the synthesis of photoactivated antibacterial and antiviral compounds
Catalog No. 50-299-1204 Supplier Cayman Chemical Supplier No. 304071 G
Add to Cart
Add to Cart
A photooxidant; upon light activation, it forms an electrophilic aromatic ketone that acts as an oxidant in organic synthesis or biological systems; has been used in the study of amino acid oxidation, as well as the synthesis of photoactivated antibacterial and antiviral compounds