Learn More
Enzo Life Sciences 4-Phenylchalcone Oxide (100mg). CAS: 82389-34-2
SDP
Catalog No. 502010554
Encompass_Preferred
Potent and selective inhibitor of microsomal and cytosolic epoxide hydrolase (IC50=68 nM). Inhibits epoxide hydrolysis of epoxyeicosatrienoic acids. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0554 Supplier Enzo Life Sciences Supplier No. BMLEI1080100
Potent and selective inhibitor of microsomal and cytosolic epoxide hydrolase (IC50=68 nM). Inhibits epoxide hydrolysis of epoxyeicosatrienoic acids. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (25mg/ml). Long Term Storage: -20°C.