Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-phenylimidazole | 670-95-1 | MFCD00005197 | 98.5% | 144.18 g/mol | C9H8N2 | 10 G
SDP

Catalog No. 5000301532
Encompass_Preferred

4-Phenylimidazole is an organic biochemical reagent (C9H8N2) used in life-science research as a building block and ligand in biochemical assays. It is supplied as a solid with high reported purity suitable for laboratory applications.

  • Purity 98.5%
  • Molecular formula C9H8N2
  • Molecular weight 144.18 g/mol
  • Physical appearance off-white to yellow solid
  • Available pack sizes 5 g, 10 g, 50 g

Catalog No. 50-003-01532 Supplier Medchemexpress LLC Supplier No. HYW01567110G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Phenylimidazole is an organic biochemical reagent (C9H8N2) used in life-science research as a building block and ligand in biochemical assays. It is supplied as a solid with high reported purity suitable for laboratory applications.

  • Purity 98.5%
  • Molecular formula C9H8N2
  • Molecular weight 144.18 g/mol
  • Physical appearance off-white to yellow solid
  • Available pack sizes 5 g, 10 g, 50 g

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.