Learn More
Inorganic Ventures 4 Phosphate Modifier
SDP
Catalog No. 50723672
Encompass_Preferred
Inorganic Ventures' 4% Phosphate Modifier is a 125mL reference material set in a water matrix for stability. This ammonium dihydrogen phosphate matrix modifier is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-672 Supplier Inorganic Ventures Supplier No. MMP40125ML
Add to Cart
Add to Cart
Inorganic Ventures' 4% Phosphate Modifier is a 125mL reference material set in a water matrix for stability. This ammonium dihydrogen phosphate matrix modifier is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search