Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-Phthalimidobutyric acid | 3130-75-4 | MFCD00196079 | 98.8% | 233.22 g/mol | C12H11NO4 | 250 MG
SDP

Catalog No. 5000209651
Encompass_Preferred

O-Phthalimide-C3-acid (4-phthalimidobutyric acid) is a small-molecule hapten containing a terminal carboxyl group and a three-carbon spacer arm. It is used to prepare hapten-protein conjugates for antibody production and antigen design.

  • Hapten with a terminal carboxyl group for activation and coupling to amine groups.
  • Suitable for conjugation to carrier proteins in antigen design.
  • Molecular formula C12H11NO4.
  • Molecular weight 233.22 g/mol.
  • Reported purity 98.79% (HPLC).
  • Available in small pack sizes such as 250 MG and 500 MG.

Catalog No. 50-002-09651 Supplier Medchemexpress LLC Supplier No. 250MGHYW046063
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

O-Phthalimide-C3-acid (4-phthalimidobutyric acid) is a small-molecule hapten containing a terminal carboxyl group and a three-carbon spacer arm. It is used to prepare hapten-protein conjugates for antibody production and antigen design.

  • Hapten with a terminal carboxyl group for activation and coupling to amine groups.
  • Suitable for conjugation to carrier proteins in antigen design.
  • Molecular formula C12H11NO4.
  • Molecular weight 233.22 g/mol.
  • Reported purity 98.79% (HPLC).
  • Available in small pack sizes such as 250 MG and 500 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.