Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-piperidinamine, N-[(1R,2S)-2-phenylcyclopropyl]-, dihydrochloride | 2102933-95-7 | 99.5% | 289.24 | C14H22Cl2N2 | 25 MG
SDP

Catalog No. 5000211819
Encompass_Preferred

GSK-LSD1 dihydrochloride is a potent, selective, irreversible inhibitor of lysine-specific demethylase 1 (LSD1) with an in vitro IC50 of 16 nM. Supplied as a high-purity crystalline solid (C14H22Cl2N2, MW 289.24), it is intended for research use in biochemical and cellular studies; follow recommended storage and safety procedures.

  • Potent LSD1 inhibition with IC50 16 nM.
  • High purity suitable for biochemical assays (99.45%).
  • Solid, white to light yellow crystalline form for consistent handling.
  • Molecular weight 289.24 and formula C14H22Cl2N2 for reference.
  • Available in convenient pack sizes including 25 MG vial.

Catalog No. 50-002-11819 Supplier Medchemexpress LLC Supplier No. HY100546A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GSK-LSD1 dihydrochloride is a potent, selective, irreversible inhibitor of lysine-specific demethylase 1 (LSD1) with an in vitro IC50 of 16 nM. Supplied as a high-purity crystalline solid (C14H22Cl2N2, MW 289.24), it is intended for research use in biochemical and cellular studies; follow recommended storage and safety procedures.

  • Potent LSD1 inhibition with IC50 16 nM.
  • High purity suitable for biochemical assays (99.45%).
  • Solid, white to light yellow crystalline form for consistent handling.
  • Molecular weight 289.24 and formula C14H22Cl2N2 for reference.
  • Available in convenient pack sizes including 25 MG vial.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.