Learn More
Medchemexpress LLC 4-piperidinamine, N-[(1R,2S)-2-phenylcyclopropyl]-, hydrochloride (1:2), rel- | 2102933-95-7 | 99.5% | 289.24 g/mol | C14H22Cl2N2 | 1 MG
SDP
Catalog No. 5000212683
Encompass_Preferred
GSK-LSD1 dihydrochloride is a potent, selective, and irreversible inhibitor of lysine specific demethylase 1 (LSD1) with an IC50 of 16 nM. It is provided for research use in epigenetics and pharmacology studies. Chemical identifiers include CAS 2102933-95-7, formula C14H22Cl2N2, molecular weight 289.24 g/mol, and reported purity ~99.45%.
  • Potent, selective, irreversible LSD1 inhibition with low nanomolar potency.
  • High chemical purity suitable for research applications.
  • Dihydrochloride salt form for improved handling and solubility.
  • Well-characterized with supporting certificate of analysis available.
  • Intended for epigenetic and pharmacological research use.
Catalog No. 50-002-12683 Supplier Medchemexpress LLC Supplier No. HY100546A1MG
GSK-LSD1 dihydrochloride is a potent, selective, and irreversible inhibitor of lysine specific demethylase 1 (LSD1) with an IC50 of 16 nM. It is provided for research use in epigenetics and pharmacology studies. Chemical identifiers include CAS 2102933-95-7, formula C14H22Cl2N2, molecular weight 289.24 g/mol, and reported purity ~99.45%.
  • Potent, selective, irreversible LSD1 inhibition with low nanomolar potency.
  • High chemical purity suitable for research applications.
  • Dihydrochloride salt form for improved handling and solubility.
  • Well-characterized with supporting certificate of analysis available.
  • Intended for epigenetic and pharmacological research use.