Learn More
Medchemexpress LLC 4-Piperidineacetic acid, 1-[[1-phenyl-3-(2-thienyl)-1H-pyrazol-5-yl]carbonyl]-, methyl ester | 1282128-68-0 | 98.0% | 409.50 | 1 ML
SDP
Catalog No. 5000384681
Encompass_Preferred
KB-208 is a phagocytosis inhibitor that improves immune thrombocytopenia (ITP) in mouse models. It is suitable for immunology research.
  • Improves immune thrombocytopenia (ITP) in mouse models
  • Does not affect other blood parameters
  • Does not elevate serum toxicity biomarkers
  • For research use only
Catalog No. 50-003-84681 Supplier Medchemexpress LLC Supplier No. HY17050610MM/1ML
KB-208 is a phagocytosis inhibitor that improves immune thrombocytopenia (ITP) in mouse models. It is suitable for immunology research.
  • Improves immune thrombocytopenia (ITP) in mouse models
  • Does not affect other blood parameters
  • Does not elevate serum toxicity biomarkers
  • For research use only