Learn More
Accela Chembio Inc 4-piperidineacetic Acid | 5g | 51052-78-9 | MFCD02093097 | 95+% | Shelf Life: 540 Days | Moisture Sensitive/nitrogen Or Argon
Supplier:  Accela Chembio Inc SY0025395G
Encompass_Preferred
4-piperidineacetic Acid | 5g | 51052-78-9 | MFCD02093097 | 95+% | Shelf Life: 540 Days | Moisture Sensitive/nitrogen Or Argon
Catalog No. 50-249-855