Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-Piperidinecarboxamide, N-hydroxy-1-(2-methoxyethyl)-4-[[4-[4-(trifluoromethoxy)phenoxy]phenyl]sulfonyl]-, hydrochloride (1:1) | 226395-93-3 | 99.4% | 554.96 | 10 MG
SDP

Catalog No. 5000345354
Encompass_Preferred

SD-2590 hydrochloride is a potent MMP inhibitor with IC50 values of <0.1, <0.1, 0.18, and 1.7 nM for MMP2, MMP13, MMP9, and MMP8, respectively. It inhibits dilation of the left ventricle in rats. This product is for research use only.

  • Potent MMP inhibitor
  • Inhibits dilation of the left ventricle in rats
  • Solid appearance
  • White to off-white color
  • Ships at room temperature
  • Store at -20°C, sealed, away from moisture
  • Soluble in DMSO at 55.5 mg/mL

Catalog No. 50-003-45354 Supplier Medchemexpress LLC Supplier No. HY1101161MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SD-2590 hydrochloride is a potent MMP inhibitor with IC50 values of <0.1, <0.1, 0.18, and 1.7 nM for MMP2, MMP13, MMP9, and MMP8, respectively. It inhibits dilation of the left ventricle in rats. This product is for research use only.

  • Potent MMP inhibitor
  • Inhibits dilation of the left ventricle in rats
  • Solid appearance
  • White to off-white color
  • Ships at room temperature
  • Store at -20°C, sealed, away from moisture
  • Soluble in DMSO at 55.5 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.