Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Piperidineethanol 96.0+%, TCI America™
SDP

Catalog No. P09925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P09925G 5 g
P099225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P09925G Supplier TCI America Supplier No. P09925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 622-26-4
Molecular Formula C7H16NO
Molecular Weight (g/mol) 130.21
MDL Number MFCD00006008
InChI Key LDSQQXKSEFZAPE-UHFFFAOYSA-O
Synonym 4-piperidineethanol, 2-piperidin-4-yl ethanol, 4-2-hydroxyethyl piperidine, 2-piperidin-4-yl ethan-1-ol, 4-piperidine ethanol, piperidine-4-ethanol, 4-ethanolpiperidine, 2-4-piperidyl ethanol, 2-piperidin-4-yl-ethanol, 4-hydroxyethyl-piperidine
PubChem CID 73953
IUPAC Name 4-(2-hydroxyethyl)piperidin-1-ium
SMILES OCCC1CC[NH2+]CC1

Specifications

Melting Point 50°C
Color White
Boiling Point 228°C
Quantity 5 g
Formula Weight 129.20
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Piperidineethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.