Learn More
Medchemexpress LLC 4-Piperidinone, 2,2,6,6-tetramethyl- | 826-36-8 | 155.24 | 10 G
SDP
Catalog No. 5000415755
Encompass_Preferred
Triacetonamine (2,2,6,6-Tetramethyl-4-piperidone) is used as an intermediate for the synthesis of pharmaceutical products, pesticides, and photostabilizers for polymers. It has oral activity and can induce acute liver failure (ALF) in rats.
  • Used as an intermediate in chemical synthesis.
  • Exhibits oral activity.
  • Can induce acute liver failure in rats.
Catalog No. 50-004-15755 Supplier Medchemexpress LLC Supplier No. HYN1131B10G
Triacetonamine (2,2,6,6-Tetramethyl-4-piperidone) is used as an intermediate for the synthesis of pharmaceutical products, pesticides, and photostabilizers for polymers. It has oral activity and can induce acute liver failure (ALF) in rats.
  • Used as an intermediate in chemical synthesis.
  • Exhibits oral activity.
  • Can induce acute liver failure in rats.