Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(4-Piperidinophenyl)methylamine, 97%, Thermo Scientific™

Catalog No. CC33913DE
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC33913DE 5 g Amber Glass Bottle
CC33913DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC33913DE Supplier Thermo Scientific Chemicals Supplier No. CC33913DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 214759-73-6
Molecular Formula C12H18N2
Molecular Weight (g/mol) 190.29
MDL Number MFCD06659080
InChI Key WROBJEHXMLQDQP-UHFFFAOYSA-N
Synonym 4-piperidinophenyl methylamine, 4-piperidin-1-yl phenyl methanamine, 4-piperidinobenzylamine, 4-piperidin-1-yl-benzylamine, 1-4-piperidin-1-yl phenyl methanamine, 4-piperidin-1yl benzylamine, 4-piperidin-1ylphenyl methylamine, 4-piperidylphenyl methylamine, 4-1-piperidyl phenyl methanamine
PubChem CID 2795551
IUPAC Name (4-piperidin-1-ylphenyl)methanamine
SMILES NCC1=CC=C(C=C1)N1CCCCC1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 190.29
Percent Purity 97%
Chemical Name or Material (4-Piperidinophenyl)methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.