Learn More
Accela Chembio Inc 4-piperidinopiperidine | 25g | 4897-50-1 | MFCD00006475 | 98% | Shelf Life: 2160 Days | Light Sensitive/nitrogen Or Argon/+4
Catalog No. 50353633
Encompass_Preferred
4-piperidinopiperidine | 25g | 4897-50-1 | MFCD00006475 | 98% | Shelf Life: 2160 Days | Light Sensitive/nitrogen Or Argon/+4
Catalog No. 50-353-633 Supplier Accela Chembio Inc Supplier No. SY00312425G
4-piperidinopiperidine | 25g | 4897-50-1 | MFCD00006475 | 98% | Shelf Life: 2160 Days | Light Sensitive/nitrogen Or Argon/+4