Learn More
Electron Microscopy Sciences Bench Top 4 Place Pipettor Holder
SDP
Catalog No. 5030591
Encompass_Preferred
The pipettor holder is perfect for use in DNA workstations, where U.V. anti-germicidal amp are used. It is resistant to most solvents and is easy to clean. It accomodates most major brands of pippetors. Supplied with non-skid feet. Made from high density polyethylene, Dimensions: 12" (L) x 8" (H), x 8" (D)
Catalog No. 50-305-91 Supplier Electron Microscopy Sciences Supplier No. 6437804
The pipettor holder is perfect for use in DNA workstations, where U.V. anti-germicidal amp are used. It is resistant to most solvents and is easy to clean. It accomodates most major brands of pippetors. Supplied with non-skid feet. Made from high density polyethylene, Dimensions: 12" (L) x 8" (H), x 8" (D)