Learn More
Charter Medical 4INPLASMA TRANSFER SET 96/CS
Catalog No. NC0788160
Encompass
Manufactured by Incell Corporation and supplied by Charter Medical:These disposable sets are designed to obtain a sample of blood or blood components and/or to perform a sterile transfer of blood or blood components from one container to another. Features and Benefits: Needle-less systems prevent accidental needle sticks. Multiple configurations available. Tubing is compatible with sterile connection technology.May be used for Quality Control sampling
Catalog No. NC0788160 Supplier Charter Medical Supplier No. 0322090
Manufactured by Incell Corporation and supplied by Charter Medical:These disposable sets are designed to obtain a sample of blood or blood components and/or to perform a sterile transfer of blood or blood components from one container to another. Features and Benefits: Needle-less systems prevent accidental needle sticks. Multiple configurations available. Tubing is compatible with sterile connection technology.May be used for Quality Control sampling