Learn More
MSC Sellars Scrim R400 Wipers, Low-Lint, Pop-Up Box, 175 Sheets/Box
Catalog No. 502819152
Encompass
These Scrim R400 Wipers are engineered for demanding cleaning tasks that require durability and precision. They feature a scrim-reinforced design for exceptional strength and tear resistance, making them suitable for industrial, automotive, and maintenance environments.
  • Scrim-reinforced design provides superior strength and tear resistance
  • Low-lint construction ensures surfaces remain clean and streak-free
  • Compact pop-up box for portable and space-saving storage and access
  • High absorbency effectively soaks up liquids and oils
  • Versatile applications for industrial, automotive, and general-purpose cleaning tasks
  • Made from up to 100% recycled tissue
Catalog No. 50-281-9152 Supplier MSC Supplier No. 15692551
These Scrim R400 Wipers are engineered for demanding cleaning tasks that require durability and precision. They feature a scrim-reinforced design for exceptional strength and tear resistance, making them suitable for industrial, automotive, and maintenance environments.
  • Scrim-reinforced design provides superior strength and tear resistance
  • Low-lint construction ensures surfaces remain clean and streak-free
  • Compact pop-up box for portable and space-saving storage and access
  • High absorbency effectively soaks up liquids and oils
  • Versatile applications for industrial, automotive, and general-purpose cleaning tasks
  • Made from up to 100% recycled tissue